4-(Methylsulfonyl)-3-nitrobenzotrifluorid

Lieferant: Apollo Scientific

PC7475-1G PC7475-5G
APOSPC7475-1GEA 96 EUR
APOSPC7475-1G APOSPC7475-5G
4-(Methylsulfonyl)-3-nitrobenzotrifluorid
4-(Methylsulfonyl)-3-nitrobenzotrifluorid
Formel: C₈H₆F₃NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00107523
CAS-Nummer: 364-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR