2-Fluoro-4-methylsulphonylphenol 95+%

Lieferant: Apollo Scientific
Gefahr

PC7486-1G PC7486-250MG PC7486-5G
APOSPC7486-1GEA 69.6 EUR
APOSPC7486-1G APOSPC7486-250MG APOSPC7486-5G
2-Fluoro-4-methylsulphonylphenol 95+%
2-Fluoro-4-methylsulphonylphenol
Formel: C₇H₇FO₃S
Molecular Weight: 190.19 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09264549
CAS-Nummer: 398456-87-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR