4-Cyano-2-(trifluoromethyl)phenylhydrazine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydrazino-3-(trifluoromethyl)benzonitrile, 4-Hydrazinyl-3-(trifluoromethyl)benzonitrile

PC7498-1G PC7498-5G
APOSPC7498-1GEA 81.2 EUR
APOSPC7498-1G APOSPC7498-5G
4-Cyano-2-(trifluoromethyl)phenylhydrazine 97%
4-Cyano-2-(trifluoromethyl)phenylhydrazine
Formel: C₈H₆F₃N₃
Molecular Weight: 201,15 g/mol
Schmelzpunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04972777
CAS-Nummer: 184163-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR