3-(Trifluoromethyl)benzaldoxime 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC7504-1G
PC7504-5G
APOSPC7504-1GEA
84.1
EUR
APOSPC7504-1G
APOSPC7504-5G
3-(Trifluoromethyl)benzaldoxime 97%
3-(Trifluoromethyl)benzaldoxime
Formel:
C₈H₆F₃NO Molecular Weight: 189.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00033235 CAS-Nummer: 368-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...