3-Amino-4-fluorphenol 99%

Lieferant: Apollo Scientific
Achtung

PC7511-25G PC7511-5G
APOSPC7511-25GEA 119 EUR
APOSPC7511-25G APOSPC7511-5G
3-Amino-4-fluorphenol 99%
3-Amino-4-fluorphenol
Formel: C₆H₆FNO
Molecular Weight: 127.1200 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09033850
CAS-Nummer: 62257-16-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR