(2-Nitro-4-(trifluormethyl)phenyl)essigsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-[2-Nitro-4-(trifluoromethyl)phenyl]acetic acid
PC7539-5G
PC7539-25G
APOSPC7539-5GEA
0
EUR
APOSPC7539-5G
APOSPC7539-25G
(2-Nitro-4-(trifluormethyl)phenyl)essigsäure 97%
(2-Nitro-4-(trifluormethyl)phenyl)essigsäure
Formel:
C₉H₆F₃NO₄ Molecular Weight: 249,15 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00091534 CAS-Nummer: 1735-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...