7-Fluoro-1-benzothiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC7541-1G
PC7541-5G
APOSPC7541-1GEA
0
EUR
APOSPC7541-1G
APOSPC7541-5G
7-Fluoro-1-benzothiophene-2-carboxylic acid
7-Fluoro-1-benzothiophene-2-carboxylic acid
Formel:
C₉H₅FO₂S Schmelzpunkt: 255…258 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07377074 CAS-Nummer: 550998-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...