1-[(4-Cyano-2-fluoro)phenyl]piperidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC7561-1G PC7561-5G
APOSPC7561-1GEA 126 EUR
APOSPC7561-1G APOSPC7561-5G
1-[(4-Cyano-2-fluoro)phenyl]piperidine-3-carboxylic acid
1-[(4-Cyano-2-fluoro)phenyl]piperidine-3-carboxylic acid
Formel: C₁₃H₁₃FN₂O₂
Molecular Weight: 248,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750918
CAS-Nummer: 1000339-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR