1-(Benzyloxy)-4-brom-2,3-difluorbenzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7574-1G
PC7574-5G
APOSPC7574-1GEA
40.6
EUR
APOSPC7574-1G
APOSPC7574-5G
1-(Benzyloxy)-4-brom-2,3-difluorbenzol 98%
1-(Benzyloxy)-4-brom-2,3-difluorbenzol
Formel:
C₁₃H₉BrF₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09475823 CAS-Nummer: 941294-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...