2,6-Difluor-3-pyridinboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Difluoropyridine-3-boronic acid

PC7582-1G PC7582-100MG PC7582-5G
APOSPC7582-1GEA 69.6 EUR
APOSPC7582-1G APOSPC7582-100MG APOSPC7582-5G
2,6-Difluor-3-pyridinboronsäure 96%
2,6-Difluoro-3-pyridineboronic acid
Formel: C₅H₄BF₂NO₂
Molecular Weight: 158,90 g/mol
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788245
CAS-Nummer: 136466-94-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR