3-(Trifluormethyl)benzoylacetonitril 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3-(Trifluormethyl)phenyl)-3-ketopropionsäurenitril, 3-(3-(Trifluormethyl)phenyl)-3-oxopropionsäurenitril

PC7586-10G PC7586-1G PC7586-5G
APOSPC7586-10GEA 219 EUR
APOSPC7586-10G APOSPC7586-1G APOSPC7586-5G
3-(Trifluormethyl)benzoylacetonitril 97%
3-(Trifluormethyl)benzoylacetonitril
Formel: C₁₀H₆F₃NO
Molecular Weight: 213,16 g/mol
Schmelzpunkt: 58…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052315
CAS-Nummer: 27328-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR