4-Brom-2-fluorbenzamid
Lieferant: Apollo Scientific
|
Achtung
|
PC7599-1G
PC7599-5G
APOSPC7599-1GEA
33.5
EUR
APOSPC7599-1G
APOSPC7599-5G
4-Brom-2-fluorbenzamid
4-Brom-2-fluorbenzamid
Formel:
C₇H₅BrFNO Molecular Weight: 218,03 g/mol Schmelzpunkt: 151…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094432 CAS-Nummer: 292621-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...