Dimethyl(methylthio)sulfoniumtetrafluorborat 97%

Lieferant: Apollo Scientific

Synonyme: DMTSF

PC7613-1G PC7613-5G
APOSPC7613-1GEA 125 EUR
APOSPC7613-1G APOSPC7613-5G
Dimethyl(methylthio)sulfoniumtetrafluorborat 97%
Dimethyl(methylthio)sulphonium tetrafluoroborate
Formel: C₃H₉BF₄S₂
Molecular Weight: 196,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011806
CAS-Nummer: 5799-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR