3-(Trifluoromethyl)cyclohexanol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Trifluoromethyl)cyclohexanol, cis/trans

PC7668G-1G PC7668G-5G
APOSPC7668G-1GEA 72.5 EUR
APOSPC7668G-1G APOSPC7668G-5G
3-(Trifluoromethyl)cyclohexanol 97%
3-(Trifluoromethyl)cyclohexanol
Formel: C₇H₁₁F₃O
Molecular Weight: 168,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00102143
CAS-Nummer: 454-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR