(α,α,α-Trifluor-4-tolyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-(Trifluormethyl)phenyl)essigsäure
PC7700-100G
PC7700-25G
PC7700-500G
APOSPC7700-100GEA
161
EUR
APOSPC7700-100G
APOSPC7700-25G
APOSPC7700-500G
(α,α,α-Trifluor-4-tolyl)essigsäure 98%
(α,α,α-Trifluor-4-tolyl)essigsäure
Formel:
(C₆H₄(CF₃))CH₂COOH Molecular Weight: 204.15 g/mol Schmelzpunkt: 79…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004352 CAS-Nummer: 32857-62-8 EINECS: 251-263-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...