4-Fluor-3-hydroxybenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7701-25G
APOSPC7701-25GEA
125
EUR
APOSPC7701-25G
4-Fluor-3-hydroxybenzoesäure 98%
4-Fluoro-3-hydroxybenzoic acid
Formel:
C₇H₅FO₃ Molecular Weight: 156,11 g/mol Schmelzpunkt: 214…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269641 CAS-Nummer: 51446-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...