8-Fluor-2-methylchinolin-7-boronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC7706-1G
PC7706-250MG
APOSPC7706-1GEA
771
EUR
APOSPC7706-1G
APOSPC7706-250MG
8-Fluor-2-methylchinolin-7-boronsäure 95%
8-Fluor-2-methylchinolin-7-boronsäure
Formel:
C₁₀H₉BFNO₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD09475865 CAS-Nummer: 957035-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...