5-Fluor-2-methoxy-3-pyridinboronsäure 98%

Lieferant: Apollo Scientific
Achtung

PC7707-250MG PC7707-1G
APOSPC7707-250MGEA 144 EUR
APOSPC7707-250MG APOSPC7707-1G
5-Fluor-2-methoxy-3-pyridinboronsäure 98%
5-Fluoro-2-methoxy-3-pyridineboronic acid
Formel: C₆H₇BFNO₃
Schmelzpunkt: 86…88 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07368239
CAS-Nummer: 957120-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR