4-(Trifluormethyl)phenylhydrazin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydrazinobenzotrifluorid, 1-(4-(Trifluormethyl)phenyl)hydrazin
PC7717-5G
PC7717-25G
PC7717-1G
APOSPC7717-5GEA
55.1
EUR
APOSPC7717-5G
APOSPC7717-25G
APOSPC7717-1G
4-(Trifluormethyl)phenylhydrazin 97%
4-(Trifluormethyl)phenylhydrazin
Formel:
C₇H₇F₃N₂ Molecular Weight: 176,14 g/mol Siedepunkt: 118…122 °C/17mmHg Schmelzpunkt: 59…66 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00042508 CAS-Nummer: 368-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...