7-Fluor-4(3H)-chinazolinon

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Fluoro-4-hydroxyquinazoline, 7-Fluoro-4-quinazolinone, 7-Fluoro-4-quinazolinol

PC7721-1G PC7721-250MG
APOSPC7721-1GEA 29 EUR
APOSPC7721-1G APOSPC7721-250MG
7-Fluor-4(3H)-chinazolinon
7-Fluoroquinazolin-4(3H)-one
Formel: C₈H₅FN₂O
Molecular Weight: 164,1400 g/mol
Schmelzpunkt: 240…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05664874
CAS-Nummer: 16499-57-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR