1-Brom-2-chlor-5-fluorbenzol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-chloro-4-fluorobenzene

PC7751-100G PC7751-25G PC7751-500G
APOSPC7751-100GEA 69.6 EUR
APOSPC7751-100G APOSPC7751-25G APOSPC7751-500G
1-Brom-2-chlor-5-fluorbenzol 98%
1-Brom-2-chlor-5-fluorbenzol
Formel: C₆H₃BrClF
Molecular Weight: 209,44 g/mol
Dichte: 1,75 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00061120
CAS-Nummer: 201849-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR