3-(Trifluormethyl)-1H-pyrazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Trifluormethyl)-1H-pyrazol
PC7766B-100G
PC7766B-25G
PC7766B-5G
APOSPC7766B-100GEA
214
EUR
APOSPC7766B-100G
APOSPC7766B-25G
APOSPC7766B-5G
3-(Trifluormethyl)-1H-pyrazol 97%
3-(Trifluoromethyl)-1H-pyrazole
Formel:
C₄H₃F₃N₂ Molecular Weight: 136,08 g/mol Siedepunkt: 70 °C/2mmHg Schmelzpunkt: 45…47 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00115018 CAS-Nummer: 20154-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...