7-(Trifluormethyl)-4-chinolinol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-7-(trifluoromethyl)quinoline
PC7767-25G
PC7767-5G
APOSPC7767-25GEA
561
EUR
APOSPC7767-25G
APOSPC7767-5G
7-(Trifluormethyl)-4-chinolinol 97%
7-(Trifluormethyl)-4-chinolinol
Formel:
C₁₀H₆F₃NO Molecular Weight: 213,1560 g/mol Schmelzpunkt: 266…269 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006779 CAS-Nummer: 322-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...