α,α,α,2,4-Pentafluortoluol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Difluoro-1-(trifluoromethyl)benzene, 2,4-Difluorobenzotrifluoride

PC7770-1G PC7770-25G PC7770-5G
APOSPC7770-1GEA 21.9 EUR
APOSPC7770-1G APOSPC7770-25G APOSPC7770-5G
α,α,α,2,4-Pentafluortoluol
α,α,α,2,4-Pentafluortoluol
Formel: C₇H₃F₅
Lagertemperatur: Raumtemperatur
MDL: MFCD04972808
CAS-Nummer: 64248-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR