Ethyl-2,3,4-trifluorbenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2,3,4-trifluorobenzoate
PC7775-100G
PC7775-25G
PC7775-5G
APOSPC7775-100GEA
122
EUR
APOSPC7775-100G
APOSPC7775-25G
APOSPC7775-5G
Ethyl-2,3,4-trifluorbenzoat
Ethyl-2,3,4-trifluorbenzoat
Formel:
C₉H₇F₃O₂ Molecular Weight: 204.1459 g/mol Siedepunkt: 205…207 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06204408 CAS-Nummer: 351354-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...