1-(3-Chlor-2-fluorphenyl)biguanid Hydrochlorid

Lieferant: Apollo Scientific
Achtung

PC7805-25G PC7805-5G
APOSPC7805-25GEA 406 EUR
APOSPC7805-25G APOSPC7805-5G
1-(3-Chlor-2-fluorphenyl)biguanid Hydrochlorid
1-(3-Chlor-2-fluorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₀Cl₂FN₅
Lagertemperatur: Raumtemperatur
MDL: MFCD03094446
CAS-Nummer: 1030829-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR