5,6-Difluorindol
Lieferant: Apollo Scientific
|
Achtung
|
PC7808-1G
PC7808-5G
APOSPC7808-1GEA
34.8
EUR
APOSPC7808-1G
APOSPC7808-5G
5,6-Difluorindol
5,6-Difluorindol
Formel:
C₈H₅F₂N Molecular Weight: 153.13 g/mol Schmelzpunkt: 99 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01075212 CAS-Nummer: 169674-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...