(2,3,4-Trifluorphenyl)essigsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2,3,4-Trifluorophenyl)acetic acid

PC7818C-1G PC7818C-5G PC7818C-10G
APOSPC7818C-1GEA 21.9 EUR
APOSPC7818C-1G APOSPC7818C-5G APOSPC7818C-10G
(2,3,4-Trifluorphenyl)essigsäure 97%
(2,3,4-Trifluorphenyl)essigsäure
Formel: C₆H₂F₃CH₂COOH
Molecular Weight: 190,12 g/mol
Schmelzpunkt: 103…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061235
CAS-Nummer: 243666-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR