(2,3,5-Trifluorphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2,3,5-Trifluorophenyl)acetic acid

PC7818D-1G PC7818D-5G PC7818D-10G
APOSPC7818D-1GEA 119 EUR
APOSPC7818D-1G APOSPC7818D-5G APOSPC7818D-10G
(2,3,5-Trifluorphenyl)essigsäure 98%
(2,3,5-Trifluorphenyl)essigsäure
Formel: C₆H₂F₃CH₂COOH
Molecular Weight: 190,12 g/mol
Schmelzpunkt: 106…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083547
CAS-Nummer: 132992-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR