(2,4,6-Trifluorphenyl)essigsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4,6-Trifluorophenylacetic acid

PC7818G-1G PC7818G-25G PC7818G-5G
APOSPC7818G-1GEA 27.7 EUR
APOSPC7818G-1G APOSPC7818G-25G APOSPC7818G-5G
(2,4,6-Trifluorphenyl)essigsäure 97%
(2,4,6-Trifluorphenyl)essigsäure
Formel: (C₆H₂F₃)CH₂COOH
Molecular Weight: 190.12 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061218
CAS-Nummer: 209991-63-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR