(2,3,4-Trifluorphenyl)acetonitril 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,3,4-Trifluorbenzylcyanid
PC7818J-1G
PC7818J-5G
PC7818J-25G
APOSPC7818J-1GEA
21.9
EUR
APOSPC7818J-1G
APOSPC7818J-5G
APOSPC7818J-25G
(2,3,4-Trifluorphenyl)acetonitril 97%
(2,3,4-Trifluorphenyl)acetonitril
Formel:
C₈H₄F₃N Molecular Weight: 171.12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061234 CAS-Nummer: 243666-13-9 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...