(2,3,6-Trifluorphenyl)acetonitril 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,3,6-Trifluorbenzylcyanid

PC7818L-1G PC7818L-5G PC7818L-25G
APOSPC7818L-1GEA 21.9 EUR
APOSPC7818L-1G APOSPC7818L-5G APOSPC7818L-25G
(2,3,6-Trifluorphenyl)acetonitril 98%
(2,3,6-Trifluorphenyl)acetonitril
Formel: C₈H₄F₃N
Molecular Weight: 171,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061219
CAS-Nummer: 114152-21-5
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR