(3,4,5-Trifluorphenyl)acetonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,4,5-Trifluorbenzylcyanid
PC7818O-1G
PC7818O-25G
PC7818O-5G
APOSPC7818O-1GEA
21.9
EUR
APOSPC7818O-1G
APOSPC7818O-25G
APOSPC7818O-5G
(3,4,5-Trifluorphenyl)acetonitril 97%
(3,4,5-Trifluorphenyl)acetonitril
Formel:
C₈H₄F₃N Molecular Weight: 171,12 g/mol Schmelzpunkt: 28…34 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00083531 CAS-Nummer: 220228-03-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...