1-(4-(Trifluormethoxy)phenyl)biguanid Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[4-(Trifluoromethoxy)phenyl]biguanide hydrochloride
PC7822-25G
PC7822-5G
PC7822-100G
APOSPC7822-25GEA
65.4
EUR
APOSPC7822-25G
APOSPC7822-5G
APOSPC7822-100G
1-(4-(Trifluormethoxy)phenyl)biguanid Hydrochlorid
1-(4-(Trifluormethoxy)phenyl)biguanid Hydrochlorid
Formel:
C₉H₁₁ClF₃N₅O Molecular Weight: 297,66 g/mol Schmelzpunkt: 212…213 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569536 CAS-Nummer: 42823-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...