2-(Difluormethoxy)benzylamin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2-(Difluoromethoxy)phenyl)methanamine

PC7845-1G PC7845-25G PC7845-5G
APOSPC7845-1GEA 23.2 EUR
APOSPC7845-1G APOSPC7845-25G APOSPC7845-5G
2-(Difluormethoxy)benzylamin 98%
2-(Difluormethoxy)benzylamin
Formel: C₈H₉F₂NO
Molecular Weight: 173.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236229
CAS-Nummer: 243863-36-7
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR