2,3,6-Trifluor-4-(trifluormethyl)pyridin 97%

Lieferant: Apollo Scientific

PC7846-1G PC7846-500MG
APOSPC7846-1GEA 400 EUR
APOSPC7846-1G APOSPC7846-500MG
2,3,6-Trifluor-4-(trifluormethyl)pyridin 97%
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine
Formel: C₆HF₆N
Molecular Weight: 201,07 g/mol
Siedepunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03840203
CAS-Nummer: 84940-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR