2,3-Difluor-4-ethoxybenzoesäure 97%

Lieferant: Apollo Scientific

PC7871-5G PC7871-1G
APOSPC7871-5GEA 159 EUR
APOSPC7871-5G APOSPC7871-1G
2,3-Difluor-4-ethoxybenzoesäure 97%
2,3-Difluoro-4-ethoxybenzoic acid
Formel: C₉H₈F₂O₃
Schmelzpunkt: 201…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02258849
CAS-Nummer: 124728-45-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR