2,4,6-Trifluorphenylhydrazin 97+%
Lieferant: Apollo Scientific
|
Achtung
|
PC7903-1G
PC7903-250MG
PC7903-100MG
APOSPC7903-1GEA
0
EUR
APOSPC7903-1G
APOSPC7903-250MG
APOSPC7903-100MG
2,4,6-Trifluorphenylhydrazin 97+%
2,4,6-Trifluorphenylhydrazin
Formel:
C₆H₅F₃N₂ Schmelzpunkt: 97…101 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03095329 CAS-Nummer: 80025-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...