Vanadiumoxytrifluorid

Lieferant: Apollo Scientific
Gefahr

Synonyme: Vanadiumoxidtrifluorid, Vanadyltrifluorid, Vanadium(V)oxyfluorid, Vanadiumoxyfluorid, Vanadiumtrifluoridoxid, Vanadium(V)trifluoridoxid, Trifluoroxovanadium

PC7960-25G PC7960-5G
APOSPC7960-25GEA 328 EUR
APOSPC7960-25G APOSPC7960-5G
Vanadiumoxytrifluorid
Vanadiumoxytrifluorid
Formel: F₃OV
Molecular Weight: 123,94 g/mol
Schmelzpunkt: 1406 °C
Dichte: 3,363 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042555
CAS-Nummer: 13709-31-4
EINECS: 237-250-7
UN: 3290
ADR: 6.1+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR