4-Cyan-2-fluorbenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
PC7963-100G
PC7963-25G
PC7963-500G
APOSPC7963-100GEA
278
EUR
APOSPC7963-100G
APOSPC7963-25G
APOSPC7963-500G
4-Cyan-2-fluorbenzoesäure
4-Cyan-2-fluorbenzoesäure
Formel:
C₈H₄FNO₂ Schmelzpunkt: 214…216 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094454 CAS-Nummer: 164149-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...