2-Iod-4-nitro-6-(trifluormethyl)anilin 95+%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-3-iodo-5-nitrobenzotrifluoride
PC7986-1G
PC7986-500MG
PC7986-5G
APOSPC7986-1GEA
212
EUR
APOSPC7986-1G
APOSPC7986-500MG
APOSPC7986-5G
2-Iod-4-nitro-6-(trifluormethyl)anilin 95+%
2-Iod-4-nitro-6-(trifluormethyl)anilin
Formel:
C₇H₄F₃IN₂O₂ Schmelzpunkt: 168…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD14560547 CAS-Nummer: 400-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...