Yttrium(III)fluorid 99.99%
Lieferant: Apollo Scientific
Synonyme:
Yttriumtrifluorid
PC8024-100G
PC8024-25G
APOSPC8024-100GEA
379
EUR
APOSPC8024-100G
APOSPC8024-25G
Yttrium(III)fluorid 99.99%
Yttrium(III)fluorid
Formel:
F₃Y Molecular Weight: 145,9 g/mol Schmelzpunkt: 1387 °C Dichte: 4,01 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049714 CAS-Nummer: 13709-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...