3,3,3-Trifluoro-DL-alanine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-3,3,3-trifluoropropanoic acid, H-DL-3,3,3-TriF-Ala-OH
PC8041-1G
PC8041-100MG
PC8041-250MG
APOSPC8041-1GEA
226
EUR
APOSPC8041-1G
APOSPC8041-100MG
APOSPC8041-250MG
3,3,3-Trifluoro-DL-alanine
3,3,3-Trifluoro-DL-alanine
Formel:
C₃H₄F₃NO₂ Molecular Weight: 143,07 g/mol Schmelzpunkt: 231…234 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004263 CAS-Nummer: 17463-43-3 EINECS: 241-478-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...