N,N-Dimethyl-4-fluor-2-nitroanilin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-N,N-dimethyl-2-nitroaniline, N,N-Dimethyl 4-fluoro-2-nitroaniline

PC8048-1G PC8048-5G
APOSPC8048-1GEA 21.9 EUR
APOSPC8048-1G APOSPC8048-5G
N,N-Dimethyl-4-fluor-2-nitroanilin
N,N-Dimethyl-4-fluor-2-nitroanilin
Formel: C₈H₉FN₂O₂
Molecular Weight: 184.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116334
CAS-Nummer: 69261-01-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR