5-(2-Fluorphenyl)-4-oxazolcarbonsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
PC8053-1G
PC8053-250MG
APOSPC8053-1GEA
478
EUR
APOSPC8053-1G
APOSPC8053-250MG
5-(2-Fluorphenyl)-4-oxazolcarbonsäure 96%
5-(2-Fluorophenyl)-4-oxazolecarboxylic acid
Formel:
C₁₀H₆FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07376192 CAS-Nummer: 347187-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...