5-Fluoro-2-(trifluoromethyl)pyridine 97%
Lieferant: Apollo Scientific
PC8069-1G
PC8069-5G
APOSPC8069-1GEA
120
EUR
APOSPC8069-1G
APOSPC8069-5G
5-Fluoro-2-(trifluoromethyl)pyridine 97%
5-Fluoro-2-(trifluoromethyl)pyridine
Formel:
C6H3F4N Molecular Weight: 165,883 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864698 CAS-Nummer: 936841-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...