3-Brom-2-fluoranilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC8089-5G
PC8089-1G
PC8089-25G
APOSPC8089-5GEA
26.1
EUR
APOSPC8089-5G
APOSPC8089-1G
APOSPC8089-25G
3-Brom-2-fluoranilin 98%
3-Brom-2-fluoranilin
Formel:
C₆H₅BrFN Molecular Weight: 190.01 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09864700 CAS-Nummer: 58534-95-5 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...