3-Chlor-4-nitrobenzotrifluorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlor-1-nitro-4-(trifluormethyl)benzol, 3-Chlor-4-nitrobenzotrifluorid, 1-Chlor-2-nitro-5-(trifluormethyl)benzol

PC8117-1G PC8117-25G PC8117-5G
APOSPC8117-1GEA 21.9 EUR
APOSPC8117-1G APOSPC8117-25G APOSPC8117-5G
3-Chlor-4-nitrobenzotrifluorid 97%
3-Chlor-4-nitrobenzotrifluorid
Formel: C₇H₃ClF₃NO₂
Molecular Weight: 225.55 g/mol
Siedepunkt: 54 °C/ 0.4mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00272564
CAS-Nummer: 402-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR