5-[(4-Bromophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%

Lieferant: Apollo Scientific

PC8125-1G PC8125-500MG PC8125-5G
APOSPC8125-1GEA 533 EUR
APOSPC8125-1G APOSPC8125-500MG APOSPC8125-5G
5-[(4-Bromophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
5-[(4-Bromophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formel: C₁₂H₈BrF₃N₂OS
Molecular Weight: 365.18 g/mol
Schmelzpunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01935942
CAS-Nummer: 321848-45-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR