1-Chloroacetyl-4-[3-chloro-5-(trifluoromethyl)pyrid-2-yl]piperazine 97%

Lieferant: Apollo Scientific

PC8138-1G PC8138-500MG PC8138-5G
APOSPC8138-1GEA 533 EUR
APOSPC8138-1G APOSPC8138-500MG APOSPC8138-5G
1-Chloroacetyl-4-[3-chloro-5-(trifluoromethyl)pyrid-2-yl]piperazine 97%
1-Chloroacetyl-4-[3-chloro-5-(trifluoromethyl)pyrid-2-yl]piperazine
Formel: C₁₂H₁₂Cl₂F₃N₃O
Molecular Weight: 342,14 g/mol
Schmelzpunkt: 84…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172868
CAS-Nummer: 260553-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR